accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2HNO

CAS RN 13162-05-5

IUPAC Name: N-ethenylformamide
Synonyms: polyvinylformamide
Molecular Weight (g/mol): 71.08
Molecular Formula: C3H5NO
InChi Key: ZQXSMRAEXCEDJD-UHFFFAOYSA-N
SMILES: C=CNC=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)