accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3

CAS RN 13166-10-4

IUPAC Name: 2-(propan-2-ylidene)propanedinitrile
Synonyms: 2-(propan-2-ylidene)propanedinitrile
Molecular Weight (g/mol): 106.13
Molecular Formula: C6H6N2
InChi Key: NVBHWAQBDJEGEO-UHFFFAOYSA-N
SMILES: CC(C)=C(C#N)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results