accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1317-34-6

IUPAC Name: oxo[(oxomanganio)oxy]manganese
Synonyms: manganese(III) oxide
Molecular Weight (g/mol): 157.87
Molecular Formula: Mn2O3
InChi Key: GEYXPJBPASPPLI-UHFFFAOYSA-N
SMILES: O=[Mn]O[Mn]=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)