accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1317-40-4

IUPAC Name: copper(2+) sulfanediide
Synonyms: CuS|copper sulphide|cupric sulfide|kupfer(II)-sulfid|copper(2+) sulfide|monocopper monosulfide
Molecular Weight (g/mol): 95.61
Molecular Formula: CuS
InChi Key: OMZSGWSJDCOLKM-UHFFFAOYSA-N
SMILES: [S--].[Cu++]
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)