CAS RN 1317-41-5
IUPAC Name:
copper(2+) selandiide
Synonyms:
copper(2+) selandiide
Molecular Weight (g/mol):
142.52
Molecular Formula:
CuSe
InChi Key:
QMULOZLYOQCZOH-UHFFFAOYSA-N
SMILES:
[Cu++].[Se--]
1
–
1
of
1
results
1
Copper(II) selenide, 99.5% (metals basis)
CAS: 1317-41-5 Molecular Formula: CuSe MDL Number: MFCD00016058