accessibility menu, dialog, popup

UserName

Viewing profile as user:
SCo

CAS RN 1317-42-6

IUPAC Name: λ²-cobalt(2+) sulfanediide
Synonyms: cobalt sulfide
Molecular Weight (g/mol): 90.99
Molecular Formula: CoS
InChi Key: INPLXZPZQSLHBR-UHFFFAOYSA-N
SMILES: [S--].[Co++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results