accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131717-99-2

IUPAC Name: bis(4-tert-butylphenyl)iodanium 4-methylbenzene-1-sulfonate
Synonyms: bis(4-tert-butylphenyl)iodanium tosylate
Molecular Weight (g/mol): 564.52
Molecular Formula: C27H33IO3S
InChi Key: UEJFJTOGXLEPIV-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)