accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOF

CAS RN 131747-60-9

IUPAC Name: (2-fluoropyridin-4-yl)methanol
Synonyms: (2-fluoropyridin-4-yl)methanol
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: IAFXOPAZVVMMLE-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)