accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3ONH3CCH3NOCH3H3CCH3

CAS RN 131833-93-7

IUPAC Name: 4-tert-butyl-2-[2-(4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
Synonyms: 4-tert-butyl-2-[2-(4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol): 294.44
Molecular Formula: C17H30N2O2
InChi Key: DPMGLJUMNRDNMX-UHFFFAOYNA-N
SMILES: CC(C)(C)C1COC(=N1)C(C)(C)C1=NC(CO1)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)