accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13184-86-6

IUPAC Name: 4-(ethoxymethyl)-2-methoxyphenol
Synonyms: 4-(ethoxymethyl)-2-methoxyphenol
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: KOCVACNWDMSLBM-UHFFFAOYSA-N
SMILES: CCOCC1=CC=C(O)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results