accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131840-54-5

IUPAC Name: N-methyl-N-(2-methylphenyl)formamide
Synonyms: N-methyl-N-(2-methylphenyl)formamide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: WWWSJNJIEHCOSE-UHFFFAOYSA-N
SMILES: CN(C=O)C1=CC=CC=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results