accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131864-67-0

IUPAC Name: 2,6-bis[4-(propan-2-yl)-4,5-dihydro-1,3-oxazol-2-yl]pyridine
Synonyms: 2,6-bis(4-isopropyl-4,5-dihydro-1,3-oxazol-2-yl)pyridine
Molecular Weight (g/mol): 301.39
Molecular Formula: C17H23N3O2
InChi Key: CSGQGLBCAHGJDR-UHFFFAOYNA-N
SMILES: CC(C)C1COC(=N1)C1=CC=CC(=N1)C1=NC(CO1)C(C)C
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results