accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOCH3

CAS RN 13192-04-6

IUPAC Name: 1,5-dimethyl 2-oxopentanedioate
Synonyms: 1,5-dimethyl 2-oxopentanedioate
Molecular Weight (g/mol): 174.15
Molecular Formula: C7H10O5
InChi Key: TXIXSLPEABAEHP-UHFFFAOYSA-N
SMILES: COC(=O)CCC(=O)C(=O)OC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)