accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 131922-07-1

IUPAC Name: 1-(1,4-dimethylpiperazin-2-yl)methanamine
Synonyms: 1-(1,4-dimethylpiperazin-2-yl)methanamine
Molecular Weight (g/mol): 143.23
Molecular Formula: C7H17N3
InChi Key: DKFODFYKOIZTMO-UHFFFAOYNA-N
SMILES: CN1CCN(C)C(CN)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results