accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13194-68-8

IUPAC Name: 4-iodo-2-methylaniline
Synonyms: 4-iodo-2-methylaniline
Molecular Weight (g/mol): 233.05
Molecular Formula: C7H8IN
InChi Key: BGKLFAQCHHCZRZ-UHFFFAOYSA-N
SMILES: CC1=CC(I)=CC=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results