accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONO

CAS RN 13200-83-4

IUPAC Name: 1-formylpyrrolidine-2-carboxylic acid
Synonyms: 1-formylpyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 143.14
Molecular Formula: C6H9NO3
InChi Key: DHDRGOURKDLAOT-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCN1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results