accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13209-15-9

IUPAC Name: 1,2-bis(dibromomethyl)benzene
Synonyms: 1,2-bis(dibromomethyl)benzene
Molecular Weight (g/mol): 421.75
Molecular Formula: C8H6Br4
InChi Key: LNAOKZKISWEZNY-UHFFFAOYSA-N
SMILES: BrC(Br)C1=CC=CC=C1C(Br)Br
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (5)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)