accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 13211-01-3

IUPAC Name: 1-{[1,1'-biphenyl]-4-yl}butan-1-one
Synonyms: 1-{[1,1'-biphenyl]-4-yl}butan-1-one
Molecular Weight (g/mol): 224.3
Molecular Formula: C16H16O
InChi Key: WOFNMZNJGZJHJO-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results