accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONH2(S)CH3H3CH3CH3CCH3

CAS RN 13211-31-9

IUPAC Name: tert-butyl (2S)-2-amino-3-methylbutanoate
Synonyms: tert-butyl (2S)-2-amino-3-methylbutanoate
Molecular Weight (g/mol): 173.26
Molecular Formula: C9H19NO2
InChi Key: RJBVJBGFJIHJSZ-ZETCQYMHSA-N
SMILES: CC(C)[C@H](N)C(=O)OC(C)(C)C
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)