accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHN

CAS RN 13220-46-7

IUPAC Name: 4-methyl-2,3-dihydro-1H-indol-2-one
Synonyms: 4-methyl-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: USRZZPHRQZGXFH-UHFFFAOYSA-N
SMILES: CC1=C2CC(=O)NC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results