accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132289-66-8

IUPAC Name: methyl 2-amino-2-(thiophen-2-yl)acetate hydrochloride
Synonyms: methyl 2-amino-2-(thiophen-2-yl)acetate hydrochloride
Molecular Weight (g/mol): 207.67
Molecular Formula: C7H10ClNO2S
InChi Key: NZEQIXANVTXFEP-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results