accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132335-44-5

IUPAC Name: 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol
Synonyms: 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol
Molecular Weight (g/mol): 185.29
Molecular Formula: C9H15NOS
InChi Key: XWCNSHMHUZCRLN-UHFFFAOYNA-N
SMILES: CN(C)CCC(O)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results