accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2(S)OOHHNO

CAS RN 132388-58-0

IUPAC Name: (2S)-2-amino-3-[(triphenylmethyl)carbamoyl]propanoic acid
Synonyms: (2S)-2-amino-3-(triphenylmethylcarbamoyl)propanoic acid
Molecular Weight (g/mol): 374.44
Molecular Formula: C23H22N2O3
InChi Key: BRRPJQYCERAMFI-GNLPSFAGNA-N
SMILES: N[C@@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)