accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNONHOOH

CAS RN 132388-69-3

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-[(triphenylmethyl)carbamoyl]butanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-(triphenylmethylcarbamoyl)butanoic acid
Molecular Weight (g/mol): 488.58
Molecular Formula: C29H32N2O5
InChi Key: YEXNCDUSVVLUFM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results