accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOClH3CCH3H3C

CAS RN 132392-26-8

IUPAC Name: 5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-sulfonyl chloride
Synonyms: 5,5,8,8-tetramethyl-6,7-dihydronaphthalene-2-sulfonyl chloride
Molecular Weight (g/mol): 286.81
Molecular Formula: C14H19ClO2S
InChi Key: BEAYCJSJSMOLFG-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=CC(=C2)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results