accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH2FFFF

CAS RN 1323966-43-3

IUPAC Name: 2-fluoro-5-methyl-4-(trifluoromethyl)benzamide
Synonyms: 2-fluoro-5-methyl-4-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 221.16
Molecular Formula: C9H7F4NO
InChi Key: YHTYNOXDHGBXMS-UHFFFAOYSA-N
SMILES: CC1=C(C=C(F)C(=C1)C(N)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results