accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NCH3HS

CAS RN 13242-44-9

IUPAC Name: hydrogen 2-(dimethylamino)ethane-1-thiol chloride
Synonyms: hydrogen captamine chloride
Molecular Weight (g/mol): 141.66
Molecular Formula: C4H12ClNS
InChi Key: NRVFDGZJTPCULU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCS
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)