accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 13252-83-0

IUPAC Name: 2-hydroxy-6-methyl-4H-chromen-4-one
Synonyms: 2-hydroxy-6-methylchromen-4-one
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: CSVFZDYJGBVNEX-UHFFFAOYSA-N
SMILES: CC1=CC=C2OC(O)=CC(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results