CAS RN 13253-44-6
IUPAC Name:
(5R)-5-[2-(methylsulfanyl)ethyl]imidazolidine-2,4-dione
Synonyms:
(5R)-5-[2-(methylsulfanyl)ethyl]imidazolidine-2,4-dione
Molecular Weight (g/mol):
174.22
Molecular Formula:
C6H10N2O2S
InChi Key:
SBKRXUMXMKBCLD-SCSAIBSYSA-N
SMILES:
CSCC[C@H]1NC(=O)NC1=O
1
–
1
of
1
results
1
5-[2-(Methylthio)ethyl]hydantoin 99.0+%, TCI America™
CAS: 13253-44-6 Molecular Formula: C6H10N2O2S Molecular Weight (g/mol): 174.218 MDL Number: MFCD00143392 InChI Key: SBKRXUMXMKBCLD-UHFFFAOYSA-N PubChem CID: 567982 IUPAC Name: 5-(2-methylsulfanylethyl)imidazolidine-2,4-dione SMILES: CSCCC1C(=O)NC(=O)N1