accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1326283-60-6

IUPAC Name: 5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 267.11
Molecular Formula: C13H19BFNO3
InChi Key: JWACNMHKJHEWAQ-UHFFFAOYSA-N
SMILES: COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results