accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132664-85-8

IUPAC Name: 1-(5-methylpyrazin-2-yl)methanamine
Synonyms: 1-(5-methylpyrazin-2-yl)methanamine
Molecular Weight (g/mol): 123.16
Molecular Formula: C6H9N3
InChi Key: MPBCUCGKHDEUDD-UHFFFAOYSA-N
SMILES: CC1=CN=C(CN)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results