accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOONH(S)CH3H3CCH3

CAS RN 132684-60-7

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3,3-dimethylbutanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: VZOHGJIGTNUNNC-GOSISDBHSA-N
SMILES: CC(C)(C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results