accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNOH

CAS RN 132710-90-8

IUPAC Name: tert-butyl 4-(3-hydroxypropyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(3-hydroxypropyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 244.34
Molecular Formula: C12H24N2O3
InChi Key: LRYRQGKGCIUVON-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CCCO)CC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)