accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNCO

CAS RN 132740-43-3

IUPAC Name: 1-fluoro-4-(isocyanatomethyl)benzene
Synonyms: 1-fluoro-4-(isocyanatomethyl)benzene
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: HHSIWJYERNCLKQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(CN=C=O)C=C1
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)