CAS RN 132741-31-2
CAS RN 132741-31-2
IUPAC Name:
(2S)-2-(3,5-difluorophenyl)-2-hydroxyacetate
Synonyms:
(2S)-2-(3,5-difluorophenyl)-2-hydroxyacetate
Molecular Weight (g/mol):
187.12
Molecular Formula:
C8H5F2O3
InChi Key:
PHMLPPFFMSRWBK-ZETCQYMHSA-M
SMILES:
O[C@H](C([O-])=O)C1=CC(F)=CC(F)=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 3,5-Difluoromandelic acid, 97%, Thermo Scientific™
CAS: 132741-31-2 Molecular Formula: C8H6F2O3 Molecular Weight (g/mol): 188.13 MDL Number: MFCD00061300 InChI Key: PHMLPPFFMSRWBK-UHFFFAOYSA-N Synonym: 3,5-difluoromandelic acid,2-3,5-difluorophenyl-2-hydroxyacetic acid,3,5-difluorophenyl hydroxy acetic acid,3,5-difluoro-mandelic acid,3,5-difluoro mandelic acid,r/s-3,5-difluoromandelic acid,racemic 3,5-difluoro-mandelic acid,3,5-difluorophenylhydroxyacetic acid,3,5-difluorophenyl hydroxyacetic acid PubChem CID: 588648 IUPAC Name: 2-(3,5-difluorophenyl)-2-hydroxyacetic acid SMILES: C1=C(C=C(C=C1F)F)C(C(=O)O)O