accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132810-10-7

IUPAC Name: 1-ethyl-4-[4-(4-fluorophenyl)-5H,6H,7H,8H,9H,10H-cycloocta[b]pyridin-2-yl]piperazine
Synonyms: blonanserin
Molecular Weight (g/mol): 367.51
Molecular Formula: C23H30FN3
InChi Key: XVGOZDAJGBALKS-UHFFFAOYSA-N
SMILES: CCN1CCN(CC1)C1=CC(C2=CC=C(F)C=C2)=C2CCCCCCC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results