accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132814-92-7

IUPAC Name: 5-hexyl-5'-{5'-hexyl-[2,2'-bithiophen]-5-yl}-2,2'-bithiophene
Synonyms: 5-hexyl-5'-{5'-hexyl-[2,2'-bithiophen]-5-yl}-2,2'-bithiophene
Molecular Weight (g/mol): 498.82
Molecular Formula: C28H34S4
InChi Key: KGJQHEPGNCWZRN-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(CCCCCC)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results