accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13283-01-7

IUPAC Name: tungsten hexachloride
Synonyms: tungsten hexachloride
Molecular Weight (g/mol): 402.59
Molecular Formula: Cl6H6W
InChi Key: XHYHOBJDYHRWHO-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.Cl.Cl.[W]
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (5)
Quantity 
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (9)
  • (4)
  • (3)
  • (5)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)