CAS RN 132839-58-8
CAS RN 132839-58-8
IUPAC Name:
2-bromo-1-nitro-4-(trifluoromethyl)benzene
Synonyms:
2-bromo-1-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
270.01
Molecular Formula:
C7H3BrF3NO2
InChi Key:
DJOLBPUSSIOMKU-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=C(Br)C=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
Thermo Scientific Chemicals 3-Bromo-4-nitrobenzotrifluoride, 95%, Thermo Scientific™
CAS: 132839-58-8 Molecular Formula: C7H3BrF3NO2 Molecular Weight (g/mol): 270.005 MDL Number: MFCD09998160 InChI Key: DJOLBPUSSIOMKU-UHFFFAOYSA-N Synonym: 2-bromo-1-nitro-4-trifluoromethyl benzene,3-bromo-4-nitrobenzotrifluoride,2-bromo-4-trifluoromethyl nitrobenzene,benzene, 2-bromo-1-nitro-4-trifluoromethyl,acmc-209bq5,ksc496a5t,2-bromo-1-nitro-4-trifluoromethyl-benzene,2-bromo-4-trifluoromethylnitrobenzene PubChem CID: 14790878 IUPAC Name: 2-bromo-1-nitro-4-(trifluoromethyl)benzene SMILES: C1=CC(=C(C=C1C(F)(F)F)Br)[N+](=O)[O-]