accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N(E)

CAS RN 13284-42-9

IUPAC Name: (2E)-pent-2-enenitrile
Synonyms: (2E)-pent-2-enenitrile
Molecular Weight (g/mol): 81.12
Molecular Formula: C5H7N
InChi Key: ISBHMJZRKAFTGE-ONEGZZNKSA-N
SMILES: CC\C=C\C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results