CAS RN 13288-57-8
IUPAC Name:
tert-butyl (2S)-2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoate hydrochloride
Synonyms:
tert-butyl (2S)-2-amino-6-[(tert-butoxycarbonyl)amino]hexanoate hydrochloride
Molecular Weight (g/mol):
338.87
Molecular Formula:
C15H31ClN2O4
InChi Key:
TZBPQINFXPIRBX-VAMKTSTMNA-N
SMILES:
Cl.CC(C)(C)OC(=O)NCCCC[C@H](N)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
Promotions Available
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, 95%
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, 95%
CAS: 13288-57-8 Molecular Formula: C15H31ClN2O4 Molecular Weight (g/mol): 338.87 MDL Number: MFCD00038899 InChI Key: TZBPQINFXPIRBX-VAMKTSTMNA-N Synonym: h-lys boc-otbu.hcl,h-lys boc-otbu hcl,ne-boc-l-lysine tert-butyl ester hydrochloride,h-l-lys boc-otbu*hcl,c15h30n2o4.hcl,pubchem18975,lys boc-otbu hcl,h-lys boc-obut hcl,h-lys boc-otbu?hcl,h-lys boc-otbu hydrochloride PubChem CID: 13819885 SMILES: Cl.CC(C)(C)OC(=O)NCCCC[C@H](N)C(=O)OC(C)(C)C