accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 132883-43-3

IUPAC Name: 2,2,2-trifluoro-N-(pyrrolidin-3-yl)acetamide hydrochloride
Synonyms: 2,2,2-trifluoro-N-(pyrrolidin-3-yl)acetamide hydrochloride
Molecular Weight (g/mol): 218.6
Molecular Formula: C6H10ClF3N2O
InChi Key: CMZSIQCZAFAEDH-UHFFFAOYNA-N
SMILES: Cl.FC(F)(F)C(=O)NC1CCNC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)