accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13289-97-9

IUPAC Name: N,N-diethylaniline boron
Synonyms: diethylaniline boron
Molecular Weight (g/mol): 160.05
Molecular Formula: C10H15BN
InChi Key: KHYAFFAGZNCWPT-UHFFFAOYSA-N
SMILES: [B].CCN(CC)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (6)