accessibility menu, dialog, popup

UserName

Viewing profile as user:
POOHHO

CAS RN 13291-46-8

IUPAC Name: 2-(diphenylphosphoroso)benzene-1,4-diol
Synonyms: 2-(diphenylphosphoroso)benzene-1,4-diol
Molecular Weight (g/mol): 310.29
Molecular Formula: C18H15O3P
InChi Key: LLOXZCFOAUCDAE-UHFFFAOYSA-N
SMILES: OC1=CC(=C(O)C=C1)P(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results