accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13296-94-1

IUPAC Name: 2-bromo-4-nitroaniline
Synonyms: benzenamine, 2-bromo-4-nitro-
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: CGPPWNTVTNCHDO-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1Br)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results