accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClSNOO

CAS RN 133-06-2

IUPAC Name: 2-[(trichloromethyl)sulfanyl]-2,3,3a,4,7,7a-hexahydro-1H-isoindole-1,3-dione
Synonyms: captan LE captane caswell no. 159 orthocide 406 orthocide 7.5 micro-check 12 buvisild K agrosol S fungus ban type II captan (substance)
Molecular Weight (g/mol): 300.58
Molecular Formula: C9H8Cl3NO2S
InChi Key: LDVVMCZRFWMZSG-UHFFFAOYNA-N
SMILES: ClC(Cl)(Cl)SN1C(=O)C2CC=CCC2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results