CAS RN 133-07-3
IUPAC Name:
2-[(trichloromethyl)sulfanyl]-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms:
folpet
Molecular Weight (g/mol):
296.55
Molecular Formula:
C9H4Cl3NO2S
InChi Key:
HKIOYBQGHSTUDB-UHFFFAOYSA-N
SMILES:
ClC(Cl)(Cl)SN1C(=O)C2=CC=CC=C2C1=O
1
–
1
of
1
results
1
Folpet, SPEX CertiPrep™
CAS: 133-07-3 Molecular Formula: C9H4Cl3NO2S Molecular Weight (g/mol): 296.55 MDL Number: MFCD00047303 InChI Key: HKIOYBQGHSTUDB-UHFFFAOYSA-N PubChem CID: 8607 ChEBI: CHEBI:82019 IUPAC Name: 2-(trichloromethylsulfanyl)isoindole-1,3-dione SMILES: ClC(Cl)(Cl)SN1C(=O)C2=CC=CC=C2C1=O