accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH(S)OH(R)OH(R)OH(R)OHOHOHSbOO

CAS RN 133-51-7

IUPAC Name: (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol; oxostibinic acid
Synonyms: meglumine antimoniate
Molecular Weight (g/mol): 365.98
Molecular Formula: C7H18NO8Sb
InChi Key: XOGYVDXPYVPAAQ-SESJOKTNSA-M
SMILES: O[Sb](=O)=O.CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results