accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133-53-9

IUPAC Name: 2,4-dichloro-3,5-dimethylphenol
Synonyms: 2,4-dichloro-3,5-dimethylphenol
Molecular Weight (g/mol): 191.05
Molecular Formula: C8H8Cl2O
InChi Key: IYOLBFFHPZOQGW-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(Cl)C(C)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results