accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 133-90-4

IUPAC Name: 3-amino-2,5-dichlorobenzoic acid
Synonyms: chloramben
Molecular Weight (g/mol): 206.02
Molecular Formula: C7H5Cl2NO2
InChi Key: HSSBORCLYSCBJR-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC(C(O)=O)=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results